Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029440.3 | NP_083716.2 | 10 - 11 | 962 | 79 | |
GenBank mRNA | AK015309.1 | - | 8 - 9 | 961 | 79 | |
AK087389.1 | - | 10 - 11 | 962 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |