Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081001.2 | NP_001074470.1 | 4 - 5 | 910 | 75 | |
NM_009765.3 | NP_033895.2 | 4 - 5 | 716 | 75 | ||
GenBank mRNA | AK145017.1 | - | 4 - 5 | 877 | 75 | |
AK148272.1 | - | 4 - 5 | 714 | 75 | ||
AK164598.1 | - | 4 - 5 | 706 | 75 | ||
U65594.1 | - | 4 - 5 | 693 | 75 | ||
U72947.1 | - | 3 - 4 | 352 | 75 | ||
U82270.1 | - | 3 - 4 | 402 | 75 | ||
U89652.1 | - | 3 - 4 | 440 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |