Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081001.2 | NP_001074470.1 | 8 - 9 | 1166 | 112 | |
NM_009765.3 | NP_033895.2 | 8 - 9 | 972 | 112 | ||
GenBank mRNA | AK148272.1 | - | 8 - 9 | 970 | 112 | |
AK164598.1 | - | 6 - 7 | 1478 | 112 | ||
U65594.1 | - | 8 - 9 | 949 | 112 | ||
U82270.1 | - | 7 - 8 | 658 | 112 | ||
U89652.1 | - | 7 - 8 | 696 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |