Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500016896.mm.1 |
TC0500001842.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016896.mm.1 |
NM_001013378.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016896.mm.1 |
NM_001013378.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016896.mm.1 |
NM_001115149.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016896.mm.1 |
NM_001115149.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016896.mm.1 |
NM_001115153.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016896.mm.1 |
NM_001115153.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016896.mm.1 |
NM_001286825.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016898.mm.1 |
TC0500001842.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016898.mm.1 |
NM_001013378.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016898.mm.1 |
NM_001013378.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016898.mm.1 |
NM_001115149.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016898.mm.1 |
NM_001115149.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016898.mm.1 |
NM_001115150.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016898.mm.1 |
NM_001115150.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016898.mm.1 |
NM_001115151.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016898.mm.1 |
NM_001115151.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016898.mm.1 |
NM_001115153.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016898.mm.1 |
NM_001115153.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016898.mm.1 |
NM_001286824.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016898.mm.1 |
NM_001286825.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016893.mm.1 |
TC0500001842.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500016893.mm.1 |
NM_001115153.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500016893.mm.1 |
NM_001115153.1 |
Refseq |