Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK030269.1 | - | 2 - 3 | 366 | 148 | |
BC125478.1 | - | 2 - 3 | 351 | 40129993 | 148 | |
BC125480.1 | - | 2 - 3 | 351 | 40129995 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |