Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK030269.1 | - | 8 - 9 | 1192 | 78 | |
BC125478.1 | - | 8 - 9 | 1177 | 40129993 | 78 | |
BC125480.1 | - | 8 - 9 | 1177 | 40129995 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |