Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pglyrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207247.4 | NP_997130.2 | 4 - 5 | 596 | 95 | |
GenBank mRNA | AY518698.1 | - | 4 - 5 | 567 | 95 | |
BC128291.1 | - | 4 - 5 | 567 | 40102537 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |