Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178387.3 | NP_848474.2 | 5 - 6 | 722 | 63 | |
GenBank mRNA | AK006916.1 | - | 4 - 5 | 400 | 63 | |
BC120503.1 | - | 2 - 3 | 174 | 63 | ||
BC120505.1 | - | 2 - 3 | 174 | 63 | ||
BC144721.1 | - | 2 - 3 | 174 | 63 | ||
BC144723.1 | - | 2 - 3 | 174 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |