Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045529.3 | NP_001038994.2 | 2 - 3 | 292 | 142 | |
GenBank mRNA | AK132226.1 | - | 2 - 3 | 148 | 142 | |
AK144885.1 | - | 2 - 3 | 151 | 142 | ||
AK172896.1 | - | 2 - 3 | 292 | 142 | ||
BC098072.1 | - | 2 - 3 | 133 | 30109159 | 142 | |
BC145705.1 | - | 2 - 3 | 241 | 40131008 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |