Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019500.4 | NP_062373.3 | 1 - 2 | 314 | 90 | |
GenBank mRNA | AF314089.2 | - | 1 - 2 | 314 | 90 | |
BC132183.1 | - | 1 - 2 | 232 | 40130460 | 90 | |
BC138497.1 | - | 1 - 2 | 233 | 8861517 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |