Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206924.2 | NP_996807.1 | 2 - 3 | 488 | 64 | |
GenBank mRNA | AB016490.1 | - | 2 - 3 | 433 | 64 | |
AK004469.1 | - | 2 - 3 | 400 | 64 | ||
AK089048.1 | - | 2 - 3 | 400 | 64 | ||
AK146337.1 | - | 2 - 3 | 489 | 64 | ||
BC008139.1 | - | 2 - 3 | 452 | 3582930 | 64 | |
BG077727.2 | - | 1 - 2 | 54 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |