Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pglyrp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165968.1 | NP_001159440.1 | 2 - 3 | 197 | 78 | |
NM_207263.2 | NP_997146.2 | 2 - 3 | 197 | 78 | ||
GenBank mRNA | AY518699.1 | - | 2 - 3 | 151 | 78 | |
BC120840.1 | - | 2 - 3 | 192 | 40129763 | 78 | |
BC144902.1 | - | 2 - 3 | 197 | 9053450 | 78 | |
BC144927.1 | - | 2 - 3 | 197 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |