Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008727.5 | NP_032753.5 | 11 - 12 | 2208 | 90 | |
GenBank mRNA | AK135008.1 | - | 2 - 3 | 151 | 90 | |
BC110659.1 | - | 11 - 12 | 2088 | 30847317 | 90 | |
BC139767.1 | - | 11 - 12 | 2067 | 90 | ||
L31932.1 | - | 11 - 12 | 1856 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |