Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293741.1 | NP_001280670.1 | 5 - 6 | 400 | 79 | |
NM_026677.4 | NP_080953.1 | 5 - 6 | 531 | 79 | ||
GenBank mRNA | AB232608.1 | - | 5 - 6 | 416 | 79 | |
AK002303.1 | - | 5 - 6 | 498 | 79 | ||
AK080805.1 | - | 5 - 6 | 401 | 79 | ||
BC027214.1 | - | 5 - 6 | 524 | 3709140 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |