Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gatad2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139304.1 | NP_647465.1 | 2 - 3 | 475 | 80 | |
GenBank mRNA | AF411837.1 | - | 2 - 3 | 475 | 80 | |
AK041594.1 | - | 3 - 4 | 532 | 80 | ||
AK141586.1 | - | 4 - 5 | 762 | 80 | ||
BC130262.1 | - | 2 - 3 | 477 | 40142858 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |