Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010252.4 | NP_034382.2 | 3 - 4 | 712 | 112 | |
GenBank mRNA | AK032128.1 | - | 3 - 4 | 397 | 112 | |
AK034085.1 | - | 3 - 4 | 300 | 112 | ||
AK034332.1 | - | 3 - 4 | 713 | 112 | ||
AK038468.1 | - | 3 - 4 | 449 | 112 | ||
AK039068.1 | - | 3 - 4 | 410 | 112 | ||
AK047752.1 | - | 3 - 4 | 462 | 112 | ||
AK049956.1 | - | 3 - 4 | 450 | 112 | ||
AK140399.1 | - | 3 - 4 | 462 | 112 | ||
AK163541.1 | - | 3 - 4 | 420 | 112 | ||
BC099939.1 | - | 3 - 4 | 421 | 30090762 | 112 | |
X55272.1 | - | 3 - 4 | 495 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |