Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fnip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162999.2 | NP_001156471.1 | 12 - 13 | 1531 | 79 | |
GenBank mRNA | AK122512.1 | - | 4 - 5 | 608 | 79 | |
BC116316.1 | - | 5 - 6 | 795 | 79 | ||
BC116317.1 | - | 5 - 6 | 795 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |