Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rxfp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212452.1 | NP_997617.1 | 14 - 15 | 1119 | 95 | |
GenBank mRNA | AK137655.1 | - | 13 - 14 | 1100 | 95 | |
AY509975.1 | - | 14 - 15 | 1119 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mus musculus Probe spans exons |