Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130457.1 | NP_001123929.1 | 1 - 2 | 101 | 90 | |
NM_134011.2 | NP_598772.1 | 1 - 2 | 101 | 90 | ||
GenBank mRNA | AK028966.1 | - | 1 - 2 | 79 | 90 | |
AK129245.1 | - | 1 - 2 | 60 | 90 | ||
AK144813.1 | - | 1 - 2 | 99 | 90 | ||
AK166760.1 | - | 1 - 2 | 99 | 90 | ||
AK168088.1 | - | 1 - 2 | 99 | 90 | ||
BC016483.1 | - | 1 - 2 | 57 | 3155271 | 90 | |
BC031155.1 | - | 1 - 2 | 72 | 4976690 | 90 | |
BC031910.1 | - | 1 - 2 | 71 | 5362943 | 90 | |
BY784127.1 | - | 1 - 2 | 102 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |