Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011716.2 | NP_035846.1 | 4 - 5 | 588 | 75 | |
GenBank mRNA | AF084482.1 | - | 4 - 5 | 521 | 75 | |
AJ011971.1 | - | 4 - 5 | 515 | 75 | ||
AK144561.1 | - | 4 - 5 | 540 | 75 | ||
AK145023.1 | - | 4 - 5 | 560 | 75 | ||
AK170180.1 | - | 4 - 5 | 543 | 75 | ||
BC046988.1 | - | 4 - 5 | 524 | 6492038 | 75 | |
CX562372.1 | - | 2 - 3 | 218 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |