Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011481.3 | NP_035611.3 | 3 - 4 | 1089 | 113 | |
GenBank mRNA | AK049582.1 | - | 3 - 4 | 350 | 113 | |
BC120633.1 | - | 3 - 4 | 1068 | 40129556 | 113 | |
BC125325.1 | - | 3 - 4 | 1065 | 40129840 | 113 | |
D49427.1 | - | 3 - 4 | 1103 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |