Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009206.2 | NP_033232.2 | 2 - 3 | 889 | 85 | |
GenBank mRNA | AF035526.1 | - | 3 - 4 | 772 | 85 | |
AK138398.1 | - | 2 - 3 | 844 | 85 | ||
AK147161.1 | - | 2 - 3 | 227 | 85 | ||
BC072588.1 | - | 2 - 3 | 872 | 85 | ||
BC090964.1 | - | 2 - 3 | 441 | 30625511 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |