Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021447.2 | NP_067422.1 | 7 - 8 | 1288 | 66 | |
GenBank mRNA | AF294790.1 | - | 7 - 8 | 1188 | 66 | |
AK019655.1 | - | 3 - 4 | 244 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |