Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011773.3 | NP_035903.2 | 4 - 5 | 753 | 129 | |
GenBank mRNA | AK144568.1 | - | 4 - 5 | 737 | 129 | |
AK165796.1 | - | 4 - 5 | 716 | 129 | ||
BC066199.1 | - | 4 - 5 | 735 | 30476359 | 129 | |
U76007.1 | - | 4 - 5 | 711 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |