Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp3a25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019792.2 | NP_062766.2 | 8 - 9 | 875 | 86 | |
NR_030782.1 | 7 - 8 | 787 | 86 | |||
GenBank mRNA | AF204959.1 | - | 8 - 9 | 874 | 86 | |
AK149541.1 | - | 7 - 8 | 788 | 86 | ||
BC028855.1 | - | 8 - 9 | 866 | 5124151 | 86 | |
Y11995.1 | - | 8 - 9 | 877 | 86 |
Gene Symbol: Cyp3a59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105160.1 | NP_001098630.1 | 8 - 9 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |