Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdr16c6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080710.2 | NP_001074179.1 | 3 - 4 | 574 | 91 | |
GenBank mRNA | AK076465.1 | - | 3 - 4 | 555 | 91 | |
BC125450.1 | - | 3 - 4 | 574 | 40129965 | 91 | |
BC125452.1 | - | 3 - 4 | 574 | 40129967 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |