Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001796.4 | NP_001001796.1 | 1 - 2 | 136 | 71 | |
GenBank mRNA | AK039633.1 | - | 1 - 2 | 137 | 71 | |
BC132613.1 | - | 1 - 2 | 105 | 40130890 | 71 | |
BC145917.1 | - | 1 - 2 | 105 | 40131220 | 71 | |
BC171926.1 | - | 1 - 2 | 105 | 9053913 | 71 | |
EU670677.1 | - | 2 - 3 | 256 | 71 | ||
EU670678.1 | - | 2 - 3 | 256 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |