Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009891.2 | NP_034021.1 | 14 - 15 | 1850 | 66 | |
GenBank mRNA | AK046406.1 | - | 14 - 15 | 1851 | 66 | |
AK047708.1 | - | 13 - 14 | 1742 | 66 | ||
BC119322.1 | - | 13 - 14 | 1612 | 8734071 | 66 | |
BC120589.1 | - | 13 - 14 | 1612 | 40129512 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |