Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogdhl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081130.1 | NP_001074599.1 | 11 - 12 | 1523 | 66 | |
GenBank mRNA | BC157970.1 | - | 10 - 11 | 1323 | 9007292 | 66 |
BC157971.1 | - | 10 - 11 | 1323 | 9007293 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |