Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Birc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163247.1 | NP_001156719.1 | 5 - 6 | 790 | 59 | |
GenBank mRNA | AK053463.1 | - | 4 - 5 | 1144 | 59 | |
BC107260.2 | - | 5 - 6 | 714 | 59 | ||
BC125013.1 | - | 5 - 6 | 714 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |