Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lingo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175516.4 | NP_780725.2 | 1 - 2 | 191 | 93 | |
GenBank mRNA | AK045639.1 | - | 1 - 2 | 192 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mus musculus Probe spans exons |