Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbl1x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020601.2 | NP_065626.1 | 11 - 12 | 1281 | 79 | |
GenBank mRNA | AK030547.1 | - | 11 - 12 | 1496 | 79 | |
AK031937.1 | - | 7 - 8 | 742 | 79 | ||
AK036996.1 | - | 8 - 9 | 3112 | 79 | ||
AK038674.1 | - | 11 - 12 | 1282 | 79 | ||
BC043105.1 | - | 11 - 12 | 1404 | 6406579 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |