Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008166.2 | NP_032192.2 | 1 - 2 | 171 | 51 | |
GenBank mRNA | D10171.1 | - | 1 - 2 | 141 | 51 | |
DN176234.1 | - | 1 - 2 | 171 | 51 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Recommended Assay Mus musculus Inventoried Probe spans exons |