Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011130.2 | NP_035260.1 | 10 - 11 | 680 | 132 | |
GenBank mRNA | AF513911.1 | - | 10 - 11 | 657 | 132 | |
AK077127.1 | - | 10 - 11 | 665 | 132 | ||
AK146745.1 | - | 10 - 11 | 669 | 132 | ||
AK151436.1 | - | 10 - 11 | 665 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |