Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027338.1 | NP_081614.1 | 6 - 7 | 539 | 113 | |
GenBank mRNA | AK150754.1 | - | 6 - 7 | 560 | 113 | |
AK151035.1 | - | 6 - 7 | 560 | 113 | ||
AK156241.1 | - | 553 | 113 | |||
AK161865.1 | - | 591 | 113 | |||
BC010811.1 | - | 6 - 7 | 539 | 4210911 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |