Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010612.2 | NP_034742.2 | 3 - 4 | 618 | 64 | |
GenBank mRNA | AK054510.1 | - | 3 - 4 | 651 | 64 | |
AK141938.1 | - | 3 - 4 | 641 | 64 | ||
BC020530.1 | - | 3 - 4 | 618 | 4238984 | 64 | |
EU884114.1 | - | 3 - 4 | 648 | 64 | ||
S53103.1 | - | 3 - 4 | 629 | 64 | ||
X59397.1 | - | 3 - 4 | 565 | 64 | ||
X70842.1 | - | 3 - 4 | 643 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |