Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghitm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199122.1 | NP_001186051.1 | 5 - 6 | 698 | 77 | |
NM_078478.5 | NP_510963.1 | 5 - 6 | 651 | 77 | ||
GenBank mRNA | AF412297.1 | - | 5 - 6 | 595 | 77 | |
AK005636.1 | - | 5 - 6 | 595 | 77 | ||
AK093887.1 | - | 5 - 6 | 627 | 77 | ||
AK134592.1 | - | 5 - 6 | 581 | 77 | ||
AK146433.1 | - | 5 - 6 | 558 | 77 | ||
AK146853.1 | - | 5 - 6 | 614 | 77 | ||
AK151488.1 | - | 5 - 6 | 628 | 77 | ||
AK158904.1 | - | 5 - 6 | 641 | 77 | ||
AK159695.1 | - | 5 - 6 | 625 | 77 | ||
AK159724.1 | - | 5 - 6 | 625 | 77 | ||
AK164122.1 | - | 5 - 6 | 596 | 77 | ||
AK167970.1 | - | 5 - 6 | 622 | 77 | ||
AK168022.1 | - | 5 - 6 | 628 | 77 | ||
AK168678.1 | - | 5 - 6 | 699 | 77 | ||
AK168800.1 | - | 5 - 6 | 607 | 77 | ||
BC008622.1 | - | 5 - 6 | 629 | 3588758 | 77 | |
BC010224.1 | - | 5 - 6 | 631 | 3500685 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |