Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paxip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018878.3 | NP_061366.2 | 13 - 14 | 2707 | 90 | |
GenBank mRNA | AF104261.1 | - | 13 - 14 | 2707 | 90 | |
AK144606.1 | - | 13 - 14 | 2569 | 90 | ||
AK148141.1 | - | 13 - 14 | 91 | |||
BC066014.1 | - | 13 - 14 | 2496 | 5704566 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |