Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dkc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030307.2 | NP_001025478.1 | 1 - 2 | 268 | 89 | |
GenBank mRNA | AK136418.1 | - | 1 - 2 | 53 | 89 | |
AK144139.1 | - | 1 - 2 | 56 | 89 | ||
AK166105.1 | - | 1 - 2 | 44 | 89 | ||
BC099966.1 | - | 1 - 2 | 108 | 6435181 | 89 | |
BY121749.1 | - | 1 - 2 | 207 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |