Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136240.1 | NP_001129712.1 | 6 - 7 | 1554 | 58 | |
NM_172264.2 | NP_758468.2 | 6 - 7 | 1352 | 58 | ||
NM_175343.5 | NP_780552.1 | 6 - 7 | 1527 | 58 | ||
GenBank mRNA | AK030900.1 | - | 6 - 7 | 1528 | 58 | |
AK052715.1 | - | 6 - 7 | 1534 | 58 | ||
AK087570.1 | - | 6 - 7 | 1463 | 58 | ||
AK164042.1 | - | 6 - 7 | 1390 | 58 | ||
BC039186.1 | - | 6 - 7 | 1352 | 3979709 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |