Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ythdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB221619.1 | - | 4 - 5 | 381 | 91 | |
AK173312.1 | - | 8 - 9 | 3452 | 91 | ||
BC022697.1 | - | 5 - 6 | 550 | 91 | ||
BC037593.1 | - | 6 - 7 | 2008 | 91 | ||
BC048817.1 | - | 5 - 6 | 535 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |