Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Best1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011913.2 | NP_036043.2 | 7 - 8 | 1083 | 126 | |
GenBank mRNA | AK006549.1 | - | 5 - 6 | 702 | 126 | |
AY450427.1 | - | 6 - 7 | 907 | 126 | ||
BC139060.1 | - | 6 - 7 | 898 | 8862082 | 126 | |
BC139061.1 | - | 6 - 7 | 898 | 8862083 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |