Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shcbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011369.2 | NP_035499.1 | 11 - 12 | 1557 | 140 | |
GenBank mRNA | AF017152.1 | - | 11 - 12 | 1557 | 140 | |
AK135226.1 | - | 3 - 4 | 2179 | 140 | ||
AK137496.1 | - | 10 - 11 | 2039 | 140 | ||
AK144966.1 | - | 11 - 12 | 1580 | 140 | ||
AK162943.1 | - | 11 - 12 | 1571 | 140 | ||
BC070455.1 | - | 11 - 12 | 1577 | 30606785 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |