Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mms22l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199467.2 | NP_955761.2 | 9 - 10 | 1562 | 82 | |
GenBank mRNA | AK089531.1 | - | 2 - 3 | 302 | 82 | |
AK144799.1 | - | 9 - 10 | 1481 | 82 | ||
AK166072.1 | - | 9 - 10 | 1573 | 82 | ||
AY512923.1 | - | 4 - 5 | 696 | 82 | ||
BC094429.1 | - | 8 - 9 | 906 | 82 | ||
BC137910.1 | - | 4 - 5 | 472 | 8860929 | 82 | |
BC145028.1 | - | 4 - 5 | 472 | 9053576 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |