Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019390.3 | NP_062263.1 | 1 - 2 | 80 | 85 | |
GenBank mRNA | BY140664.1 | - | 1 - 2 | 67 | 85 | |
D14850.1 | - | 1 - 2 | 80 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus 1700 Inventoried Probe spans exons |