Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubqln4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033526.2 | NP_277068.1 | 1 - 2 | 202 | 89 | |
GenBank mRNA | AB040050.1 | - | 1 - 2 | 202 | 89 | |
AK150211.1 | - | 1 - 2 | 140 | 89 | ||
AK156515.1 | - | 1 - 2 | 155 | 89 | ||
AK160578.1 | - | 1 - 2 | 154 | 89 | ||
BC017686.1 | - | 1 - 2 | 164 | 4236887 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |