Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubqln4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033526.2 | NP_277068.1 | 8 - 9 | 1427 | 61 | |
GenBank mRNA | AB040050.1 | - | 8 - 9 | 1427 | 61 | |
AK150211.1 | - | 8 - 9 | 1364 | 61 | ||
AK156515.1 | - | 8 - 9 | 1381 | 61 | ||
AK216643.1 | - | 1 - 2 | 33 | 61 | ||
AK218439.1 | - | 1 - 2 | 33 | 61 | ||
BC017686.1 | - | 8 - 9 | 1389 | 4236887 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |