Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178246.3 | NP_839977.2 | 4 - 5 | 618 | 82 | |
GenBank mRNA | AK129286.1 | - | 4 - 5 | 597 | 82 | |
AK143911.1 | - | 4 - 5 | 619 | 82 | ||
AK155787.1 | - | 4 - 5 | 607 | 82 | ||
AK172217.1 | - | 3 - 4 | 496 | 82 | ||
BC100408.1 | - | 4 - 5 | 567 | 30734268 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |