Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025448.3 | NP_079724.1 | 2 - 3 | 457 | 66 | |
GenBank mRNA | AK005340.1 | - | 3 - 4 | 328 | 66 | |
AK010565.1 | - | 3 - 4 | 295 | 66 | ||
AK012281.1 | - | 2 - 3 | 285 | 66 | ||
AK020145.1 | - | 3 - 4 | 278 | 66 | ||
AK050505.1 | - | 2 - 3 | 458 | 66 | ||
AK084530.1 | - | 3 - 4 | 295 | 66 | ||
BC010214.1 | - | 2 - 3 | 441 | 3497894 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |