Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoa1bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144897.3 | NP_659146.1 | 2 - 3 | 278 | 95 | |
GenBank mRNA | AJ344092.1 | - | 2 - 3 | 278 | 95 | |
AK131615.1 | - | 2 - 3 | 489 | 95 | ||
AK159846.1 | - | 2 - 3 | 280 | 95 | ||
AY566271.1 | - | 2 - 3 | 278 | 95 | ||
BC058362.1 | - | 2 - 3 | 276 | 6819459 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |